Product Details

SNP ID
rs35088527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1758862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCGTCGCCGGGTCGATAGCTTC[C/G]GCCGTCTTCAAATATTGTTTAAATT
Phenotype
MIM: 611294
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ONECUT3 PubMed Links

Gene Details

Gene
ONECUT3
Gene Name
one cut homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080488.1 Intron NP_001073957.1

View Full Product Details