Product Details

SNP ID
rs28570988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3481467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACTCCAGCCTGGGCAACAGAAC[A/G]AGACCTTGTCTCTCAAAGCAAATTT
Phenotype
MIM: 611262
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOHH PubMed Links

Gene Details

Gene
DOHH
Gene Name
deoxyhypusine hydroxylase/monooxygenase
There are no transcripts associated with this gene.

Gene
SMIM24
Gene Name
small integral membrane protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136503.1 Intron NP_001129975.1

View Full Product Details