Product Details

SNP ID
rs28638429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4292877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGTATAGGACGTTGCTGTAGA[A/G]TGCATTTCCTAGGATGGATGACACA
Phenotype
MIM: 610481 MIM: 614715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHD PubMed Links

Gene Details

Gene
SHD
Gene Name
Src homology 2 domain containing transforming protein D
There are no transcripts associated with this gene.

Gene
TMIGD2
Gene Name
transmembrane and immunoglobulin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169126.1 257 Missense Mutation CTC,TTC L187F NP_001162597.1
NM_001308232.1 257 Missense Mutation CTC,TTC L71F NP_001295161.1
NM_144615.2 257 Missense Mutation CTC,TTC L191F NP_653216.2
XM_017026284.1 257 Intron XP_016881773.1

View Full Product Details