Product Details

SNP ID
rs11557525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:537034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGACCATTCTCCTGAGTGTGAT[A/C]TCCCTCCTGAACGAGCCCAACACCT
Phenotype
MIM: 116948 MIM: 600311
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC34 PubMed Links

Gene Details

Gene
CDC34
Gene Name
cell division cycle 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004359.1 583 Silent Mutation ATA,ATC I128I NP_004350.1
XM_005259690.3 583 Silent Mutation ATA,ATC I128I XP_005259747.1
XM_006722952.2 583 Silent Mutation ATA,ATC I128I XP_006723015.1
Gene
GZMM
Gene Name
granzyme M
There are no transcripts associated with this gene.

View Full Product Details