Product Details

SNP ID
rs28536599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20660876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGATTTGCGAGAAAAGGAAAAGGG[A/G]GTAGAAGAAGAGGTAGGGTTGATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372319 PubMed Links

Gene Details

Gene
LOC105372319
Gene Name
putative zinc finger protein 56
There are no transcripts associated with this gene.

Gene
ZNF626
Gene Name
zinc finger protein 626
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076675.2 Intron NP_001070143.1
NM_145297.3 Intron NP_660340.1

View Full Product Details