Product Details

SNP ID
rs34472780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9841985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATAGTGAGGAGGGCATGGGTCT[A/G]GGTGTGGTGGGCGAGGGTGTCTCGC
Phenotype
MIM: 601052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIN1 PubMed Links

Gene Details

Gene
PIN1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006221.3 Intron NP_006212.1
XM_011528068.2 Intron XP_011526370.1

View Full Product Details