Product Details

SNP ID
rs34394365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37204861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTGAGGTGTGCTGAAGATCCA[A/G]GTCTCCAAAAGTCTGGTTCCTGTGC
Phenotype
MIM: 614910 MIM: 182453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF6 PubMed Links

Gene Details

Gene
C1QTNF6
Gene Name
C1q and tumor necrosis factor related protein 6
There are no transcripts associated with this gene.

Gene
SSTR3
Gene Name
somatostatin receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001051.4 3024 UTR 3 NP_001042.1
NM_001278687.2 3024 UTR 3 NP_001265616.1
XM_005261721.4 3024 UTR 3 XP_005261778.1
XM_006724311.2 3024 UTR 3 XP_006724374.1
XM_011530349.2 3024 UTR 3 XP_011528651.1
XM_017028923.1 3024 UTR 3 XP_016884412.1
XM_017028924.1 3024 UTR 3 XP_016884413.1

View Full Product Details