Product Details

SNP ID
rs28378166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20444481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGGACCTGTTAAAAGCCAGAT[C/T]GCCAAGACCCACCCTCAGGGCTTCC
Phenotype
MIM: 613619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL22 PubMed Links

Gene Details

Gene
KLHL22
Gene Name
kelch like family member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032775.3 Intron NP_116164.2
XM_017029018.1 Intron XP_016884507.1
XM_017029019.1 Intron XP_016884508.1
XM_017029020.1 Intron XP_016884509.1
XM_017029021.1 Intron XP_016884510.1
XM_017029022.1 Intron XP_016884511.1
XM_017029023.1 Intron XP_016884512.1
XM_017029024.1 Intron XP_016884513.1
XM_017029025.1 Intron XP_016884514.1
Gene
SCARF2
Gene Name
scavenger receptor class F member 2
There are no transcripts associated with this gene.

View Full Product Details