Product Details

SNP ID
rs28685261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43167168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTGGAATCCTTTTCTGTCTGG[C/G]GCTCCACCGCCCACAATCAGCCCCA
Phenotype
MIM: 109610
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSPO PubMed Links

Gene Details

Gene
TSPO
Gene Name
translocator protein
There are no transcripts associated with this gene.

Gene
TTLL12
Gene Name
tubulin tyrosine ligase like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015140.3 2837 UTR 3 NP_055955.1

View Full Product Details