Product Details

SNP ID
rs28375915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49960607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCGCCGCACCCGGCCCTCGCCC[A/G]CCCGAAGACAGGCGCCAAGCTGCCC
Phenotype
MIM: 610580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6821 PubMed Links

Gene Details

Gene
MIR6821
Gene Name
microRNA 6821
There are no transcripts associated with this gene.

Gene
PIM3
Gene Name
Pim-3 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001852.3 113 UTR 5 NP_001001852.2

View Full Product Details