Product Details

SNP ID
rs36081692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100844302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTTCCCTGTTTTGAGAACACA[C/G]ATGACAGACATAAGACAGTGCAGTG
Phenotype
MIM: 300907 MIM: 300225
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSTF2 PubMed Links

Gene Details

Gene
CSTF2
Gene Name
cleavage stimulation factor subunit 2
There are no transcripts associated with this gene.

Gene
NOX1
Gene Name
NADPH oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271815.1 Intron NP_001258744.1
NM_007052.4 Intron NP_008983.2
NM_013955.2 Intron NP_039249.1
XM_017029407.1 Intron XP_016884896.1

View Full Product Details