Product Details

SNP ID
rs28653713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:64188508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCAACTGCCAAGAAGCCCTGTCA[G/T]TTGATGATGCTAAGGACTCGGCTAA
Phenotype
MIM: 300647
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 5052 UTR 3 NP_689637.3

View Full Product Details