Product Details

SNP ID
rs7891507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154439448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCAGCACTTTGGGAGGCCTAGG[C/T]GGGTGGATCACGAGGTCAGGAGTTC
Phenotype
MIM: 300197 MIM: 300453 MIM: 300104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6AP1 PubMed Links

Gene Details

Gene
ATP6AP1
Gene Name
ATPase H+ transporting accessory protein 1
There are no transcripts associated with this gene.

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
There are no transcripts associated with this gene.

Gene
GDI1
Gene Name
GDP dissociation inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001493.2 Intron NP_001484.1

View Full Product Details