Product Details

SNP ID
rs28614024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119575637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCTGTGTAAGGCAGTCACTTTT[A/T]AAAAGGAGCAATCATAGCACTGGTA
Phenotype
MIM: 312180
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2A PubMed Links

Gene Details

Gene
UBE2A
Gene Name
ubiquitin conjugating enzyme E2 A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282161.1 Intron NP_001269090.1
NM_003336.3 Intron NP_003327.2
NM_181762.2 Intron NP_861427.1

View Full Product Details