Product Details

SNP ID
rs35249440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:18985450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCATATTGACCCCTTTTTTGTC[C/T]TCCAACCCACTGGCAGTAGCATCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD24P4 PubMed Links

Gene Details

Gene
CD24P4
Gene Name
CD24 molecule pseudogene 4
There are no transcripts associated with this gene.

Gene
TTTY14
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details