Product Details
- SNP ID
-
rs35036633
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.Y:19459305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCTCACCACAGACAGAGCTGCC[A/G]TAGAACTCCCAAGAGCTATTCAGCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCORP1
PubMed Links
Gene Details
- Gene
- BCORP1
- Gene Name
- BCL6 corepressor pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details