Product Details

SNP ID
rs1131326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19710451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGGACCCGGAGTTCAGTCAA[A/G]GTCAGCTGTGGAGTGTCCATCCTAT
Phenotype
MIM: 426000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 3204 Silent Mutation NP_001140177.1
NM_001146706.1 3204 Silent Mutation NP_001140178.1
NM_004653.4 3204 Silent Mutation NP_004644.2
XM_005262560.2 3204 Silent Mutation XP_005262617.1
XM_005262561.2 3204 Silent Mutation XP_005262618.1
XM_011531468.2 3204 Silent Mutation XP_011529770.1

View Full Product Details