Product Details

SNP ID
rs28665421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:78903103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTTTCCCACCAAGGTATTGTA[C/T]TGTATTGTATGCCATACAATACCTT
Phenotype
MIM: 614577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMP2K PubMed Links

Gene Details

Gene
BMP2K
Gene Name
BMP2 inducible kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017593.3 Intron NP_060063.2
NM_198892.1 Intron NP_942595.1
XM_005263117.1 Intron XP_005263174.1
XM_011532102.1 Intron XP_011530404.1
XM_017008381.1 Intron XP_016863870.1
XM_017008382.1 Intron XP_016863871.1
Gene
PAQR3
Gene Name
progestin and adipoQ receptor family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040202.1 Intron NP_001035292.1
XM_005262769.4 Intron XP_005262826.1
XM_017007785.1 Intron XP_016863274.1

View Full Product Details