Product Details
- SNP ID
-
rs17882114
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122617924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTATTGAGAAGGAGAGTGAAAAG[A/T]TTAACCTTGATTTAGGCAGCCTCAG
- Phenotype
-
MIM: 605384
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78956800] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL21
- Gene Name
- interleukin 21
- Gene
- IL21-AS1
- Gene Name
- IL21 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details