Product Details

SNP ID
rs28371599
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:207323729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTAACTTTTTATTTTTATCCA[C/G]AATAGAGGCAGTAGGATGAAGTACA
Phenotype
MIM: 125240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD55 PubMed Links

Gene Details

Gene
CD55
Gene Name
CD55 molecule (Cromer blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000574.4 Intron NP_000565.1
NM_001114752.2 Intron NP_001108224.1
NM_001300902.1 Intron NP_001287831.1
NM_001300903.1 Intron NP_001287832.1
NM_001300904.1 Intron NP_001287833.1
XM_017000467.1 Intron XP_016855956.1

View Full Product Details