Product Details

SNP ID
rs17883094
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:12170024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGGGGTACCTGCTGGCCCTGG[A/G]GGCTCTTCCCTCCTTCCACCACGTC
Phenotype
MIM: 191191
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7846 PubMed Links

Gene Details

Gene
MIR7846
Gene Name
microRNA 7846
There are no transcripts associated with this gene.

Gene
TNFRSF1B
Gene Name
TNF receptor superfamily member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001066.2 Intron NP_001057.1
XM_011542060.2 Intron XP_011540362.1
XM_011542063.2 Intron XP_011540365.1
XM_017002211.1 Intron XP_016857700.1
XM_017002212.1 Intron XP_016857701.1
XM_017002213.1 Intron XP_016857702.1
XM_017002214.1 Intron XP_016857703.1
XM_017002215.1 Intron XP_016857704.1

View Full Product Details