Product Details

SNP ID
rs28364045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27307487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCCAACTCTGGGGTGGGGGAAA[A/G]GGGTCAACGTTTTCGCAGCCCCAAA
Phenotype
MIM: 137960 MIM: 606474 MIM: 600945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPV17 PubMed Links

Gene Details

Gene
MPV17
Gene Name
MPV17, mitochondrial inner membrane protein
There are no transcripts associated with this gene.

Gene
TRIM54
Gene Name
tripartite motif containing 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032546.3 708 Intron NP_115935.3
NM_187841.2 708 Intron NP_912730.2
Gene
UCN
Gene Name
urocortin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003353.3 708 UTR 3 NP_003344.1

View Full Product Details