Product Details

SNP ID
rs34691009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10032865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAACCACTTTCCAAAAAGACAA[A/G]GAAATCTCATATTGCTAATGAAGTT
Phenotype
MIM: 613984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIDECP PubMed Links

Gene Details

Gene
CIDECP
Gene Name
cell death-inducing DFFA-like effector c pseudogene
There are no transcripts associated with this gene.

Gene
FANCD2
Gene Name
Fanconi anemia complementation group D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018115.2 290 Missense Mutation AAG,AGG K33R NP_001018125.1
NM_001319984.1 290 Missense Mutation AAG,AGG K33R NP_001306913.1
NM_033084.4 290 Missense Mutation AAG,AGG K33R NP_149075.2

View Full Product Details