Product Details

SNP ID
rs28564401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27121812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGCAGGAGATGTAATGTGGCC[A/G]TACAAAGGGGTCTTCTGCAGAGCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEK10 PubMed Links

Gene Details

Gene
NEK10
Gene Name
NIMA related kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031741.3 Intron NP_001026911.1
NM_001304384.1 Intron NP_001291313.1
NM_152534.4 Intron NP_689747.3
NM_199347.3 Intron NP_955379.2
XM_006712997.3 Intron XP_006713060.1
XM_006712998.2 Intron XP_006713061.1
XM_006712999.3 Intron XP_006713062.1
XM_006713001.3 Intron XP_006713064.1
XM_006713002.1 Intron XP_006713065.1
XM_011533413.2 Intron XP_011531715.1
XM_011533414.2 Intron XP_011531716.1
XM_011533415.1 Intron XP_011531717.1
XM_017005761.1 Intron XP_016861250.1
XM_017005762.1 Intron XP_016861251.1
XM_017005763.1 Intron XP_016861252.1
XM_017005764.1 Intron XP_016861253.1
XM_017005765.1 Intron XP_016861254.1
XM_017005766.1 Intron XP_016861255.1
XM_017005767.1 Intron XP_016861256.1
XM_017005768.1 Intron XP_016861257.1
XM_017005769.1 Intron XP_016861258.1
XM_017005770.1 Intron XP_016861259.1
XM_017005771.1 Intron XP_016861260.1
XM_017005772.1 Intron XP_016861261.1
XM_017005773.1 Intron XP_016861262.1
XM_017005774.1 Intron XP_016861263.1

View Full Product Details