Product Details

SNP ID
rs6808835
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46408373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGCCCTTCTGGGCTCATGCTGG[G/T]GGCGATCCCATGTGTAAAATTCTCA
Phenotype
MIM: 608379
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCRL2 PubMed Links

Gene Details

Gene
CCRL2
Gene Name
C-C motif chemokine receptor like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130910.1 579 Silent Mutation GGG,GGT G110G NP_001124382.1
NM_003965.4 579 Silent Mutation GGG,GGT G98G NP_003956.2
XM_011534208.1 579 Silent Mutation GGG,GGT G98G XP_011532510.1
XM_011534209.1 579 Silent Mutation GGG,GGT G98G XP_011532511.1
XM_017007436.1 579 Silent Mutation GGG,GGT G98G XP_016862925.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details