Product Details

SNP ID
rs1139896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49828629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTAATTTTATTTTAC[A/G]TGAAAATCTTCCATCCTCTCCATAC
Phenotype
MIM: 605958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAIP PubMed Links

Gene Details

Gene
TRAIP
Gene Name
TRAF interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005879.2 1718 UTR 3 NP_005870.2
XM_011533264.1 1718 UTR 3 XP_011531566.1
XM_017005526.1 1718 UTR 3 XP_016861015.1

View Full Product Details