Product Details

SNP ID
rs28415187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157747291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAACTGGTGTGTTCAAGCTCA[C/T]GTCTTGACATTATAAAGCTGTAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM11 PubMed Links

Gene Details

Gene
LSM11
Gene Name
LSM11, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173491.3 Intron NP_775762.1
Gene
THG1L
Gene Name
tRNA-histidine guanylyltransferase 1 like
There are no transcripts associated with this gene.

View Full Product Details