Product Details
- SNP ID
-
rs28712194
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:1830027 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCAGTGTGCAGGCTCCGAAGTG[C/T]TCTCTGCCAGCCGTCAGAGGGCTGC
- Phenotype
-
MIM: 608136
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78406831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF10
- Gene Name
- Rho guanine nucleotide exchange factor 10
View Full Product Details