Product Details

SNP ID
rs1803069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52711481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACCCTGGGAATGTTTCTTATT[C/T]TGAGATTGGAGGGCTATCAGAACAG
Phenotype
MIM: 602708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMC6 PubMed Links

Gene Details

Gene
PSMC6
Gene Name
proteasome 26S subunit, ATPase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002806.3 Intron NP_002797.3
XM_017021472.1 Intron XP_016876961.1

View Full Product Details