Product Details

SNP ID
rs35987281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96259369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAAATTCATGAACAACTTTAT[C/G]ACAAGTTTGTAGTTCAGTAAATCTG
Phenotype
MIM: 600337
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BDKRB1 PubMed Links

Gene Details

Gene
BDKRB1
Gene Name
bradykinin receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000710.3 Intron NP_000701.2

View Full Product Details