Product Details
- SNP ID
-
rs35538974
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:102139571 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGCTCATGACACCAGCCCCGCCGC[A/G]GTTCCCCCTGACCGGCTTTCCGCTG
- Phenotype
-
MIM: 140571
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSP90AA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116641992] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSP90AA1
- Gene Name
- heat shock protein 90 alpha family class A member 1
- Gene
- WDR20
- Gene Name
- WD repeat domain 20
View Full Product Details