Product Details
- SNP ID
-
rs28482150
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:105422120 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGGTGTGCTCGGAGCTGTCAGGC[C/T]CCCCACGTGCCCGCCCCGAGGACTT
- Phenotype
-
MIM: 603526
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100507437
PubMed Links
Gene Details
- Gene
- LOC100507437
- Gene Name
- uncharacterized LOC100507437
There are no transcripts associated with this gene.
- Gene
- MTA1
- Gene Name
- metastasis associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001203258.1 |
173 |
Intron |
|
|
NP_001190187.1 |
NM_004689.3 |
173 |
Intron |
|
|
NP_004680.2 |
XM_011537305.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535607.1 |
XM_011537306.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535608.1 |
XM_011537307.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535609.1 |
XM_011537308.2 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535610.1 |
XM_011537309.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535611.1 |
XM_011537310.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_011535612.1 |
XM_017021752.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_016877241.1 |
XM_017021753.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_016877242.1 |
XM_017021754.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_016877243.1 |
XM_017021755.1 |
173 |
Missense Mutation |
CCC,CTC |
P53L |
XP_016877244.1 |
XM_017021756.1 |
173 |
Intron |
|
|
XP_016877245.1 |
XM_017021757.1 |
173 |
Intron |
|
|
XP_016877246.1 |
XM_017021758.1 |
173 |
Intron |
|
|
XP_016877247.1 |
XM_017021759.1 |
173 |
Intron |
|
|
XP_016877248.1 |
- Gene
- TEX22
- Gene Name
- testis expressed 22
There are no transcripts associated with this gene.
View Full Product Details