Product Details

SNP ID
rs28605199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:115567816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCCCTTGAGTTGTGATATTCA[C/T]ACACACACGAGTATATATACACTTA
Phenotype
MIM: 300131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLS3 PubMed Links

Gene Details

Gene
PLS3
Gene Name
plastin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136025.4 Intron NP_001129497.1
NM_001172335.2 Intron NP_001165806.1
NM_001282337.1 Intron NP_001269266.1
NM_001282338.1 Intron NP_001269267.1
NM_005032.6 Intron NP_005023.2
XM_011537534.1 Intron XP_011535836.1
Gene
PLS3-AS1
Gene Name
PLS3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details