Product Details

SNP ID
rs35100640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49744640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTGCTCTAGAGACTTTGTAG[C/T]GCTTAAATCTTGGGGATAGCTGAGC
Phenotype
MIM: 600748
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMBIM6 PubMed Links

Gene Details

Gene
TMBIM6
Gene Name
transmembrane BAX inhibitor motif containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098576.1 Intron NP_001092046.1
NM_003217.2 Intron NP_003208.2
XM_005269126.3 Intron XP_005269183.1

View Full Product Details