Product Details

SNP ID
rs35034027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86655004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCCCTCCTGGCCACACACAGGG[C/G]CTTTGACAGGGAGATAGAAAAGGTC
Phenotype
MIM: 605906 MIM: 606665
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDB3 PubMed Links

Gene Details

Gene
LDB3
Gene Name
LIM domain binding 3
There are no transcripts associated with this gene.

Gene
OPN4
Gene Name
opsin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030015.2 Intron NP_001025186.1
NM_033282.3 Intron NP_150598.1
XM_017016955.1 Intron XP_016872444.1
XM_017016956.1 Intron XP_016872445.1
XM_017016957.1 Intron XP_016872446.1

View Full Product Details