Product Details

SNP ID
rs28673918
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:40509247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTCCCTGAAGGCAAACGCCCG[C/G]GGAGCCCACAAGTCCGGGCCGCACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXO5 PubMed Links

Gene Details

Gene
EXO5
Gene Name
exonuclease 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022774.1 512 Intron NP_073611.1
XM_005271125.3 512 Intron XP_005271182.1
XM_005271126.3 512 Intron XP_005271183.1
XM_005271128.3 512 Intron XP_005271185.1
XM_011541970.2 512 Intron XP_011540272.1
XM_017002099.1 512 UTR 5 XP_016857588.1
XM_017002100.1 512 Intron XP_016857589.1
XM_017002101.1 512 Intron XP_016857590.1
XM_017002102.1 512 UTR 5 XP_016857591.1
XM_017002103.1 512 Intron XP_016857592.1
XM_017002104.1 512 Intron XP_016857593.1

View Full Product Details