Product Details

SNP ID
rs17884489
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:44806281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGTGCAGCCCTGCGGCCAGCG[A/C]CGCCTCCAGGGCACGCTGTGCACGC
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 531 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 531 Missense Mutation GCG,TCG A130S NP_001013654.1
XM_006710614.3 531 Missense Mutation GCG,TCG A188S XP_006710677.1
XM_011541375.2 531 Missense Mutation GCG,TCG A130S XP_011539677.1
XM_011541376.2 531 Missense Mutation GCG,TCG A130S XP_011539678.1
XM_017001187.1 531 Missense Mutation GCG,TCG A171S XP_016856676.1

View Full Product Details