Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198961.1 | Intron | NP_001185890.1 | |||
NM_001198962.1 | Intron | NP_001185891.1 | |||
NM_001319958.1 | Intron | NP_001306887.1 | |||
NM_018281.3 | Intron | NP_060751.2 | |||
XM_011541709.2 | Intron | XP_011540011.1 | |||
XM_011541710.2 | Intron | XP_011540012.1 | |||
XM_011541711.2 | Intron | XP_011540013.1 | |||
XM_011541712.2 | Intron | XP_011540014.1 | |||
XM_011541713.2 | Intron | XP_011540015.1 | |||
XM_011541715.2 | Intron | XP_011540017.1 | |||
XM_011541719.2 | Intron | XP_011540021.1 | |||
XM_011541720.2 | Intron | XP_011540022.1 | |||
XM_011541721.2 | Intron | XP_011540023.1 | |||
XM_011541722.2 | Intron | XP_011540024.1 | |||
XM_011541723.2 | Intron | XP_011540025.1 | |||
XM_011541724.2 | Intron | XP_011540026.1 | |||
XM_011541725.2 | Intron | XP_011540027.1 | |||
XM_011541726.2 | Intron | XP_011540028.1 | |||
XM_011541727.2 | Intron | XP_011540029.1 | |||
XM_017001638.1 | Intron | XP_016857127.1 | |||
XM_017001639.1 | Intron | XP_016857128.1 | |||
XM_017001640.1 | Intron | XP_016857129.1 | |||
XM_017001641.1 | Intron | XP_016857130.1 | |||
XM_017001642.1 | Intron | XP_016857131.1 |