Product Details

SNP ID
rs11544978
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:67685519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGACTTTGGAGGAATTCTCGGCT[A/G]GAGAGCAGAAGACCGAAAGGTGAGT
Phenotype
MIM: 126335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GADD45A PubMed Links

Gene Details

Gene
GADD45A
Gene Name
growth arrest and DNA damage inducible alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199741.1 343 Missense Mutation AGA,GGA R9G NP_001186670.1
NM_001199742.1 343 Missense Mutation AGA,GGA R9G NP_001186671.1
NM_001924.3 343 Missense Mutation AGA,GGA R9G NP_001915.1

View Full Product Details