Product Details

SNP ID
rs36071273
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:77955366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAAAACAAAAAACAGAATTAA[A/G]GTTTTTAAAAGGCAATTTTGGCCGT
Phenotype
MIM: 603444 MIM: 613121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUBP1 PubMed Links

Gene Details

Gene
FUBP1
Gene Name
far upstream element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303433.1 Intron NP_001290362.1
NM_003902.4 Intron NP_003893.2
XM_011542391.2 Intron XP_011540693.1
XM_011542392.2 Intron XP_011540694.1
XM_011542393.2 Intron XP_011540695.1
XM_017002739.1 Intron XP_016858228.1
XM_017002740.1 Intron XP_016858229.1
XM_017002741.1 Intron XP_016858230.1
XM_017002742.1 Intron XP_016858231.1
XM_017002743.1 Intron XP_016858232.1
XM_017002744.1 Intron XP_016858233.1
Gene
NEXN
Gene Name
nexilin F-actin binding protein
There are no transcripts associated with this gene.

View Full Product Details