Product Details

SNP ID
rs28507893
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:39034457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAAAGGAAGGAAAGTACACCCC[C/T]CCACCTCACCACATTGGCAAGGGGG
Phenotype
MIM: 603847
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFS5 PubMed Links

Gene Details

Gene
NDUFS5
Gene Name
NADH:ubiquinone oxidoreductase subunit S5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184979.1 Intron NP_001171908.1
NM_004552.2 Intron NP_004543.1

View Full Product Details