Product Details
- SNP ID
-
rs28383613
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:3006138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTGATCCCCCAGCCTTCTGCTC[C/G]ATCTACGGGGAAAACTGGGGAAGGC
- Phenotype
-
MIM: 160998
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NQO2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs576144322] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NQO2
- Gene Name
- NAD(P)H quinone dehydrogenase 2
View Full Product Details