Product Details

SNP ID
rs1133244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3155722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATCCACCAGGATGGCCCGAGG[A/T]ACATATTTGTTACCTGCAGGAAATA
Phenotype
MIM: 603156 MIM: 615101
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BPHL PubMed Links

Gene Details

Gene
BPHL
Gene Name
biphenyl hydrolase-like (serine hydrolase)
There are no transcripts associated with this gene.

Gene
TUBB2A
Gene Name
tubulin beta 2A class IIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001069.2 266 Silent Mutation GTA,GTT V60V NP_001060.1
NM_001310315.1 266 UTR 5 NP_001297244.1

View Full Product Details