Product Details

SNP ID
rs28720785
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1056597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTGTATTTTAAAAAATTCAG[A/T]TGTCATCTTAGTATTTTCAGAGTAT
Phenotype
MIM: 612041
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
RNF212 PubMed Links

Gene Details

Gene
RNF212
Gene Name
ring finger protein 212
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131034.3 Intron NP_001124506.1
NM_001193318.2 Intron NP_001180247.1
NM_194439.4 Intron NP_919420.1
XM_005272274.2 Intron XP_005272331.1
XM_011513439.1 Intron XP_011511741.1
XM_011513443.1 Intron XP_011511745.1
XM_011513444.1 Intron XP_011511746.1
XM_011513445.2 Intron XP_011511747.1
XM_011513446.1 Intron XP_011511748.1
XM_017008039.1 Intron XP_016863528.1
XM_017008040.1 Intron XP_016863529.1
XM_017008041.1 Intron XP_016863530.1
XM_017008042.1 Intron XP_016863531.1

View Full Product Details