Product Details

SNP ID
rs28672144
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1495672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGCCACCCGTCTTCTTGGGTC[C/G]TCGTCCCCTGCCACCCTCTGGTGCC
Phenotype
MIM: 613442 MIM: 611140
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PTX4 PubMed Links

Gene Details

Gene
PTX4
Gene Name
pentraxin 4
There are no transcripts associated with this gene.

Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 Intron NP_057195.2
XM_011522773.2 Intron XP_011521075.1
XM_011522774.2 Intron XP_011521076.1
XM_011522775.2 Intron XP_011521077.1
XM_011522776.2 Intron XP_011521078.1
XM_011522777.2 Intron XP_011521079.1
XM_011522778.2 Intron XP_011521080.1
XM_017023914.1 Intron XP_016879403.1

View Full Product Details