Product Details
- SNP ID
-
rs28707754
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70035147 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTGCACATTCTGCACATGTATC[C/T]CTGAACTTAAATTAAAATAAATCAT
- Phenotype
-
MIM: 142702
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HTN3
PubMed Links
Gene Details
- Gene
- HTN3
- Gene Name
- histatin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000200.2 |
|
Intron |
|
|
NP_000191.1 |
View Full Product Details