Product Details

SNP ID
rs28558794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8582555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTCAGGGTATTTTTCAGAACAT[G/T]GACTATTTTGTGTGATCATGATGTC
Phenotype
MIM: 608990 MIM: 601480
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS10 PubMed Links

Gene Details

Gene
ADAMTS10
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282352.1 Intron NP_001269281.1
NM_030957.3 Intron NP_112219.3
XM_006722917.3 Intron XP_006722980.1
XM_017027338.1 Intron XP_016882827.1
XM_017027339.1 Intron XP_016882828.1
XM_017027340.1 Intron XP_016882829.1
Gene
MYO1F
Gene Name
myosin IF
There are no transcripts associated with this gene.

View Full Product Details