Product Details

SNP ID
rs11537783
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48576335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCTCTGGATGGTCGTCAGATC[C/T]GTGTGGATCATGCAGGCAAGTCTGC
Phenotype
MIM: 300027
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RBM3 PubMed Links

Gene Details

Gene
RBM3
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006743.4 466 Missense Mutation CGT,TGT R78C NP_006734.1

View Full Product Details