Product Details

SNP ID
rs7885035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48963504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCATGGCATTTATTGAGCACCT[A/G]CTGGGTCCCTGGCACTGTGCTAAGT
Phenotype
MIM: 300281 MIM: 300713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCND1 PubMed Links

Gene Details

Gene
KCND1
Gene Name
potassium voltage-gated channel subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004979.5 Intron NP_004970.3
XM_011543910.2 Intron XP_011542212.1
XM_017029508.1 Intron XP_016884997.1
XM_017029509.1 Intron XP_016884998.1
XM_017029510.1 Intron XP_016884999.1
Gene
OTUD5
Gene Name
OTU deubiquitinase 5
There are no transcripts associated with this gene.

View Full Product Details