Product Details

SNP ID
rs5917603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38573942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACTGTTGATATTGACTTTGATC[A/G]CCGGGGTGAGATAATTTGTCAGGTT
Phenotype
MIM: 300096
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN7 PubMed Links

Gene Details

Gene
TSPAN7
Gene Name
tetraspanin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004615.3 Intron NP_004606.2

View Full Product Details